Order Entry
United States
ContactUsLinkComponent
 
Anti-CA9 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 303123]
  220017-364
Anti-CA9 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 303123]
  220017-364
 :  FAB2188A
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Carbonic Anhydrase 9
  • Antigen Symbol:
    CA9
  • Clonality:
    Monoclonal
  • Clone:
    303123
  • Conjugation:
    APC (Allophycocyanin)
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Size:
    100 Tests
  • Environmentally Preferable:
  • Form:
    Supplied in a saline solution containing BSA and Sodium Azide.
  • Gene ID:
    768
  • Antigen Synonyms:
    CAIX|carbonic dehydratase|membrane antigen MN|carbonic anhydrase 9|RCC-associated protein G250|MN|carbonate dehydratase IX|CA-IX|RCC-associated antigen G250|P54/58N|pMW1|renal cell carcinoma-associated antigen G250
  • Amino Acid Number:
    Pro59 to Asp414
  • Storage Temperature:
    Protect from light. Do not freeze.12 months from date of receipt, 2 to 8 °C as supplied.
  • Shipping Temperature:
    The product is shipped with polar packs. Upon receipt, store it immediately at the temperature recommended below.
  • Immunogen:
    Mouse myeloma cell line NS0-derived recombinant human Carbonic Anhydrase IX Pro59-Asp414; Accession # Q16790
  • Purification:
    Protein A or G purified from hybridoma culture supernatant
  • Cat. No.:
    220017-364
  • Supplier no.:
    FAB2188A

 

 

Type: Primary
Antigen: CA9
Clonality: Monoclonal
Clone: 303123
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human