To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Fibroblast Growth Factor 9
- Antigen Symbol:FGF9
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Environmentally Preferable:
- Form:Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
- Gene ID:2254
- Antigen Synonyms:HBFG-9|FGF-9|GAF|HBGF-9|SYNS3
- Amino Acid Number:Met1 to Ser208
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
- Shipping Temperature:The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below.
- Immunogen:S. frugiperda insect ovarian cell line Sf 21-derived recombinant human FGF-9 (R&D Systems, Catalog # 273-F9) Met1-Ser208; Accession # P31371
- Purification:Antigen Affinity-purified
- Cat. No.:220013-599
- Supplier no.:BAF273
Type: Primary
Antigen Symbol: FGF9
Antigen Name: fibroblast growth factor 9
Antigen Synonyms: FGF-9, GAF, HBFG-9, HBGF-9, SYNS3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human