To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:GDNF Family Receptor Alpha-like
- Antigen Symbol:GFRAL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
- Gene ID:404194
- Antigen Synonyms:GRAL|C6orf144|bA360D14.1|UNQ9356
- Amino Acid Number:Gln20 to Gly349
- Storage Temperature:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under s
- Shipping Temperature:The product is shipped at ambient temperature. Upon receipt, store it immediately at the temperature recommended below. *Small pack size (SP) is shipped with polar packs. Upon receipt, store it immediately at -20 to -70 °C
- Immunogen:Chinese hamster ovary cell line CHO-derived recombinant mouse GFR alpha-like isoform A Gln20-Gly349; Accession # Q6SJE0
- Purification:Antigen Affinity-purified
- Cat. No.:220012-241
- Supplier no.:AF5728
Specifications
About this item
Type: Primary
Antigen Symbol: GFRAL
Antigen Name: GDNF family receptor alpha like
Antigen Synonyms: AY457637, Gral
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse