Anti-RORC Mouse Monoclonal Antibody [clone: H6437]
Catalog # 220022-071
Supplier: R&D Systems
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Retinoid related orphan receptor gamma
- Antigen Symbol:RORC
- Clonality:Monoclonal
- Clone:H6437
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Gene ID:6097
- Antigen Synonyms:RZRG|NR1F3|RORG|RZR-GAMMA|TOR
- Storage Temperature:Store the unopened product at -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
- Immunogen:Recombinant human ROR gamma/RORC/NR1F3
- Purification:Protein A or G purified from hybridoma culture supernatant
- Cat. No.:220022-071
- Supplier no.:PP-H6437-00
Specifications
About this item
Type: Primary
Antigen: RORC
Clonality: Monoclonal
Clone: H6437
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human