Order Entry
United States
ContactUsLinkComponent
MRO Plus™ Heavy Pad, Brady
MRO Plus™ Heavy Pad, Brady
Catalog # 10809-776
Supplier:  Brady Worldwide
undefined
MRO Plus™ Heavy Pad, Brady
Catalog # 10809-776
Supplier:  Brady Worldwide
Supplier Number:  107720

Specifications

  • Description:
    Pad
  • Environmentally Preferable:
  • Capacity:
    26 gal.
  • Dimensions:
    15W" x 19L"
  • Cat. No.:
    10809-776
  • Supplier no.:
    MRO100

Specifications

About this item

The MRO Plus™ Heavy Pad absorbs all industrial liquids (water, petroleum-based and chemical-based fluids)

3-ply construction makes it stronger, more durable and reduces lint. Gray color masks drips and spills, dimpled, perfed so you can take only what you need and reduce waste, full sized, heavy weight pad. For use with oil and water-based fluids. Polypropylene material. Gray color.