ContactUsLinkComponent
Pierce™ Firefly Signal Enhancer (100X), Thermo Scientific
Pierce™ Firefly Signal Enhancer (100X), Thermo Scientific
Catalog # PI16180
Supplier:  Invitrogen
CAS Number:  
undefined
Pierce™ Firefly Signal Enhancer (100X), Thermo Scientific
Catalog # PI16180
Supplier:  Invitrogen
Supplier Number:  16180
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Pierce™ Firefly Signal Enhancer
  • Size:
    0.5 mL
  • Environmentally Preferable:
  • Cat. No.:
    PI16180
  • Supplier no.:
    16180
Specifications
About this item
Thermo Scientific Pierce Firefly Signal Enhancer is designed to complement the Pierce Firefly Luciferase Flash and Glow Assay and is recommended when maximum sensitivity is desired.
  • Up to 5-fold increase in Firefly Flash signal intensity
  • Up to 2-fold increase in Firefly Glow signal intensity and duration
  • Convenient and easy-to-use 100X solution
Caution: For Research Use Only. Not for use in diagnostic procedures.