Order Entry
United States
ContactUsLinkComponent
Ferric Ammonium Sulfate 5% in aqueous solution
Ferric Ammonium Sulfate 5% in aqueous solution
Catalog # 101410-654
Minority or Woman-Owned Business Enterprise
CAS Number:  
undefined
Ferric Ammonium Sulfate 5% in aqueous solution
Catalog # 101410-654
Supplier Number:  15505-50
Minority or Woman-Owned Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    500 ml
  • Environmentally Preferable:
  • Cat. No.:
    101410-654
  • Supplier no.:
    15505-50

Specifications

About this item

Synonyms: Alum iron, Ammonium iron sulphate dodecahydrate, Ferric ammonium sulphate dodecahydrate, Ammonium iron(III) sulphate dodecahydrate, Iron alum, Ammonium ferric sulphate dodecahydrate
 :
 :482.2 g/mol
CAS Number:7783-83-7
Formula:NH₄Fe(SO₄)₂·12H₂O
MDL Number:MFCD00150004
Boiling Pt:230
Merck Index:12,00549