Verhoeff’s Van-Gieson’s (VVG Method) is for staining elastic fibers, nuclei, collagen & other tissue elements
Results - Elastic Fibers: Blue-black to Black–Fine elastic fibrils may not be stained with this method, Nuclei: Blue to Black, Collagen: Red