To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:BCL2-Associated Athanogene 3
- Antigen Symbol:BAG3
- Clonality:Monoclonal
- Clone:5A8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:200 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:Liquid
- Antigen Synonyms:BAG-family molecular chaperone regulator-3|Bcl-2-binding protein|MGC104307|docking protein CAIR-1|CAIR-1|BCL2-associated athanogene 3|BAG-3|DKFZp434E0610|BCL2-binding athanogene 3|BAG3|BIS
- Storage Buffer:In ascites fluid
- Storage Temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:BAG3 (AAH14656.1, 1 a.a. ~ 575 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Cat. No.:10622-568
- Supplier no.:H00009531-M01A
Mouse monoclonal antibody raised against a full-length recombinant BAG3.
Type: Primary
Antigen: BAG3
Clonality: Monoclonal
Clone: 5A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human