To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Proto-Cadherin gamma subfamily C3
- Antigen Symbol:PCDHGC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:PCDH gamma C3|Protocadherin 43|Protocadherin 2|Protocadherin43|PC 43|PCDH2|PCDHGC 3|PC43|PCDH 2|Protocadherin gamma subfamily C 3|PCDGK_HUMAN.|Cadherin like 2|Protocadherin gamma C3
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10250-180
- Supplier no.:BS-11159R
Protocadherins are a large family of cadherin-like cell adhesion proteins that are involved in the establishment and maintenance of neuronal connections in the brain. There are three protocadherin gene clusters, designated alpha, beta and gamma, all of which contain multiple tandemly arranged genes. PCDH2 (protocadherin-2), also known as PCDHGC3 (protocadherin gamma subfamily C, 3) or PC43, is a 934 amino acid single-pass type I membrane protein that contains six cadherin domains and belongs to the protocadherin gamma family. Functioning as a calcium-dependent cell-adhesion protein, PCDH2 is thought to be involved in the establishment and maintenance of neuronal connections within the brain. Multiple isoforms of PCDH2 exist due to alternative splicing events.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: PCDHGC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat