Order Entry
United States
ContactUsLinkComponent
 
Anti-DHRS7 Rabbit Polyclonal Antibody (Cy3®)
  10480-548
 :  Bioss
Anti-DHRS7 Rabbit Polyclonal Antibody (Cy3®)
  10480-548
 :  Bioss
 :  BS-8262R-CY3
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Dehydrogenase/reductase (SDR family) Member 7
  • Antigen Symbol:
    DHRS7
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy3®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µL
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    51635
  • Antigen Synonyms:
    DHRS7_HUMAN.|CGI86|Retinal short chain dehydrogenase/reductase 4|RETSDR4|DHRS7A|Dehydrogenase/reductase SDR family member 7|CGI 86|retDSR4
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage Temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10480-548
  • Supplier no.:
    BS-8262R-CY3

 

 

DHRS7 (dehydrogenase/reductase (SDR family) member 7), also known as SDR34C1, CGI-86 or retSDR4, is a 339 amino acid member of the SDR family. Like other members of the SDR family, DHRS7 contains a cofactor-binding Rossman-fold domain and is thought to catalyze the oxidation and reduction of a variety of substrates such as steroids and retinoids. DHRS7 exists as two alternatively spliced isoforms that are encoded by a gene located on human chromosome 14, which houses over 700 genes and comprises nearly 3.5% of the human genome. Chromosome 14 encodes the presinilin 1 (PSEN1) gene, which is one of the three key genes associated with the development of Alzheimer's disease (AD). The SERPINA1 gene is also located on chromosome 14 and, when defective, leads to the genetic disorder ?-antitrypsin deficiency, which is characterized by severe lung complications and liver dysfunction.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DHRS7
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat