To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:General Transcription Factor 2H1
- Antigen Symbol:GTF2H1
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:2965
- Antigen Synonyms:Basic transcription factor 2 62 kDa subunit|P62|BTF2|Basic transcription factor 62 kDa subunit|TFB1|BTF2 p62|General transcription factor IIH polypeptide 1|gtf2h1|General transcription factor IIH subunit 1|BTF 2|TFIIH|TFIIH basal transcription factor complex p62 subunit|General transcription factor IIH,polypeptide 1,62kDa|TF2H1
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Immunogen:230-280/548
- Purification:Purified by Protein A
- Cat. No.:10478-500
- Supplier no.:BS-8175R-CY5
Specifications
About this item
Component of the core-TFIIH basal transcription factor involved in nucleotide excision repair (NER) of DNA and, when complexed to CAK, in RNA transcription by RNA polymerase II.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GTF2H1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat