To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:RAS (RAD and GEM)-like GTP-binding 1
- Antigen Symbol:REM1
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:28954
- Antigen Synonyms:GD:REM|GES
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage Temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10463-406
- Supplier no.:BS-7502R-HRP
REM (Rad and Gem related GTP binding protein) is a member of the Rad/Gem/Kir subfamily of Ras-like GTPases and shares with other members of this subfamily some unusual structural features. Among these are nonconservative amino acid substitutions within guanine nucleotide binding and hydrolysis domains, unique effector domains, extended N- and C-termini, and a conserved C-terminal sequence thought to mediate membrane association but lacking a classical isoprenylation motif. REM, with a predicted molecular weight of 32.9 kDa, is most highly expressed in cardiac muscle and is expressed at more moderate levels in lung, kidney and skeletal muscle. REM is phosphorylated in vivo and has been shown to interact with several 14-3-3 isoforms. It has been reported that the GTP-bound form of a related Ras-like GTPase, GEM/kir, inhibits high-voltage activated Ca2+ channel activities by interacting directly with the ?subunit. The reduced channel activities are the result of a decreased a-subunit expression at the plasma membrane. This inhibition of L-type Ca2+ channels prevents Ca2+-triggered exocytosis in hormone-secreting cells. There are data that suggest that REM similarly regulates Ca2+ channel expression.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: REM1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat