Order Entry
United States
ContactUsLinkComponent
Anti-HAS1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-HAS1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 10348-880
Supplier:  Bioss
Anti-HAS1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 10348-880
Supplier:  Bioss
Supplier Number:  BS-2946R-HRP
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Hyaluronan Synthase 1
  • Antigen Symbol:
    HAS1
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µL
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    3036
  • Antigen Synonyms:
    HA synthase 1|HuHAS1|Hyaluronate synthase 1|HAS|Hyaluronan synthase 1|Hyaluronic acid synthase 1|HAS1
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
  • Storage Temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Immunogen:
    562-578/578
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10348-880
  • Supplier no.:
    BS-2946R-HRP

Specifications

About this item

Catalyzes the addition of GlcNAc or GlcUA monosaccharides to the nascent hyaluronan polymer. Therefore, it is essential to hyaluronan synthesis a major component of most extracellular matrices that has a structural role in tissues architectures and regulates cell adhesion, migration and differentiation. This is one of the isozymes catalyzing that reaction. Also able to catalyze the synthesis of chito-oligosaccharide depending on the substrate (By similarity).

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: HAS1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat