To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Golgi-associated, Gamma Adaptin Ear Containing, ARF binding Protein 1
- Antigen Symbol:GGA1
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:26088
- Antigen Synonyms:gamma ear-containing|ADP-ribosylation factor binding protein GGA1|Gamma-adaptin related protein 1|ARF-binding protein 1|Golgi-localized
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10296-490
- Supplier no.:BS-13343R-CY5
Specifications
About this item
The GGA family of proteins (Golgi-localized, ARF-binding proteins) are ubiquitous coat proteins that facilitate the trafficking of soluble proteins from the trans-Golgi network (TGN) to endosomes/lysosomes by means of interactions with TGN-sorting receptors, ARF (ADP-ribosylation factor), and clathrin (1?). Members of the GGA family, GGA1,GGA2 (also known as VEAR) and GGA3, are multidomain proteins that bind mannose 6-phosphate receptors (MPRs) (1,2,4). GGAs have modular structures with an N-terminal VHS (VPS-27, Hrs, and STAM) domain followed by a GAT (GGA and TOM1) domain, a connecting hinge segment, and a C-terminal GAE (?adaptin ear) domain (5). The amino-terminal VHS domains of GGAs form complexes with the cytoplasmic domains of sorting receptors by recognizing acidic-cluster di-leucine (ACLL) sequences (3). GGA1 and GGA2 do not associate with each other, but they do colocalize on perinuclear membranes (2). The cytosolic domain of memapsin 2, but not that of memapsin 1, binds the VHS domains of GGA1 and GGA2 (6). The human GGA1 gene maps to chromosome 22 and encodes a protein that shares 45% sequence identity with GGA2 and GGA3 (1).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GGA1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat