Order Entry
United States
ContactUsLinkComponent
Anti-GALNS Rabbit Polyclonal Antibody (Cy7®)
Anti-GALNS Rabbit Polyclonal Antibody (Cy7®)
Catalog # 10295-354
Supplier:  Bioss
Anti-GALNS Rabbit Polyclonal Antibody (Cy7®)
Catalog # 10295-354
Supplier:  Bioss
Supplier Number:  BS-13269R-CY7
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Galactosamine-6 Sulfatase
  • Antigen Symbol:
    GALNS
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy7®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Size:
    100 µL
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    2588
  • Antigen Synonyms:
    GALNAC6S|GAS|GalN6S|MPS4A
  • Storage Buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage Temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping Temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Cat. No.:
    10295-354
  • Supplier no.:
    BS-13269R-CY7

Specifications

About this item

Chondroitinase is a 522 amino acid protein that localizes to the lysosome and functions as an exohydrolase that is essential for the degradation of glycosaminoglycans, keratan sulfate and chondroitin 6-sulfate. Using calcium as a cofactor, Chondroitinase, which exists as a disulfide linked oligomer, catalyzes the hydrolysis of the 6-sulfate group on target substrates. Defects in the gene encoding Chondroitinase are the cause of mucopolysaccharidosis type 4A (MPS4A), an autosomal recessive lysosomal storage disease that is characterized by the intracellular accumulation of keratan sulfate and chondroitin-6-sulfate and is associated with dental anomalies, short stature and, in some cases, death in the second or third decade of life.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GALNS
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat