To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Farnesyl Diphosphate Synthase
- Antigen Symbol:FDPS
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Gene ID:2224
- Antigen Synonyms:FPPS|FPS
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10293-340
- Supplier no.:BS-13153R-A350
Specifications
About this item
FDPS is a 419 amino acid enzyme belonging to the FPP/GGPP synthetase family. Localized to cytoplasm and peroxisome, FDPS expression is regulated by phorbol esters and polyunsaturated fatty acids. FDPS assists in cholesterol biosynthesis, post-translational protein modifications and synthesis of steroid hormones in the isoprenoid pathway.FDPS catalyzes the formation of farnesyl diphosphate (FPP), a precursor for several classes of essential metabolites including sterols, dolichols, carotenoids, and ubiquinones. FDPS is inactivated by interferon-induced RSAD2, which may result in the disruption of lipid rafts at the plasma membrane. Existing as a homodimer, FDPS may have anti-viral effects when inactivated by RSAD2. Reduced activity of FDPS in liver may partly be the cause of Zellweger syndrome and neonatal adrenoleukodystrophy, both of which are known to be peroxisomal deficiency diseases.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: FDPS
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat