To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Lymphocyte Antigen 6 Complex Locus C1
- Antigen Symbol:LY6C
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:Ly6c2|Ly6c|AA959465|CD59|Ly-6C|AA682074
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10278-538
- Supplier no.:BS-12327R-A488
The gene encoding the mouse alloantigen, Ly-6C, maps to chromosome 15 and encodes a 131 amino acid protein that belongs to the Ly-6 family of glycosyl-phosphatidylinositol (GPI)-linked proteins. Ly-6 family members share amino acid homology throughout a distinctive cysteine rich protein domain that incorporates O-linked carbohydrates. Murine Ly-6 molecules have unique patterns of tissue expression during hematopoiesis from multipotential stem cells to lineage committed precursor cells, and on specific leukocyte subpopulations in the peripheral lymphoid tissues. Ly-6C is predominantly expressed on murine peripheral CD8 T cells. Ly-6C is involved in endothelial adhesion, the killing of target cells by CTLs, inducing TCR-mediated activation of IL-2 and IFN-?production in CD8 T cells and the homing of CD8 T cells. In addition, Ly-6C may act as a signaling molecule of LFA-1 activation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LY6C
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human