To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Peroxin 5R/PEX5-related Protein
- Antigen Symbol:PEX5L
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10270-696
- Supplier no.:BS-12096R-A555
Specifications
About this item
Peroxisomes are single-membrane bound organelles present in virtually all eukaryotic cells. They are involved in numerous catabolic and anabolic pathways, including beta-oxidation of very long chain fatty acids, metabolism of hydrogen peroxide, plasmalogen biosynthesis and bile acid synthesis. The Peroxin gene family, which includes more than 20 members, is required for peroxisome biogenesis. Peroxin 5R, also known as PEX5-related protein or Peroxisome biogenesis factor 5-like, is a 626 amino acid protein that is mainly expressed in brain, with some expression in testis and pancreas. Peroxin 5R contains five TPR repeats, which enable protein-protein interactions and assembly of large multiprotein complexes. There are three isoforms of Peroxin 5R that are produced as a result of alternative splicing events. These isoforms bind C-terminal peroxisome-targeting signals in a similar manner to Peroxin-5. Peroxin 5R interacts with Rab 8b, possibly playing a role in vesicular trafficking and neurotransmitter release.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Peroxin 5R/PEX5-related protein
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat