To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Neural Proliferation, Differentiation And Control, 1
- Antigen Symbol:NPDC1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:NPDC1|NPDC1_HUMAN.|Neural proliferation dferentiation and control protein 1|CAB1|AI314472|NPDC-1
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10267-636
- Supplier no.:BS-11913R-A350
NPDC-1 (Neural Proliferation Differentiation and Control-1) is expressed in neurons once they have stopped dividing and begun to differentiate. NPDC-1 is transported from the Golgi apparatus via vesicles before becoming internalized by endosomes at the cell membrane. NPDC-1 interacts with Cdk2, D-type cyclins, and the transcription factor E2F1. This interaction can lead to an increased replication time, and might have implications in final neural differentiation and apoptosis. NPDC-1 has been shown to colocalize with synaptic vesicle proteins: synaptophysin, synaptobrevin 2, and Rab3 GEP (Rab3 GTP/GDP exchange protein). One function of NPDC-1 is to regulate retinoic acid-mediated events by directly interacting with retinoid receptors. The amino acid sequence of NPDC-1 is highly conserved between mouse, rat, and human.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NPDC1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat