To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Lin-7 homolog C
- Antigen Symbol:LIN7C
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:MALS3|LIN-7-C|LIN-7C|VELI3|MALS-3
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10267-256
- Supplier no.:BS-11872R-CY7
Specifications
About this item
Velis are a family of small synaptic proteins that interact with other proteins at the post-synaptic density (PSD) of neuronal synapses. Velis contain the PDZ motif involved in recruiting cell adhesion molecules, receptors, and channels. Veli1 (also designated Lin-7A and MALS-1), Veli2 (also designated Lin-7B and MALS-2), and Veli3 (also designated Lin-7C and MALS-3) are mammalian homologs of C. elegans LIN-7. Veli proteins are ubiquitously expressed with high expression in brain, liver, and testis. Velis are localized at the synaptic junctions in neurons. Velis bind to CASK, a neurexin-binding protein highly concentrated in synapses, and Mint1, a binding partner with a vesicle trafficking protein.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LIN7C
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat