To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Neuron Navigator 1
- Antigen Symbol:NAV1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Form:liquid
- Antigen Synonyms:Steerin 1|Protein Steerin 1|POMFIL3|Protein Steerin1|Nav1|Steerin1|mNav1|unc-53 homolog 1|NAV1_HUMAN.|FLJ14203|DKFZp781D0314|Pore membrane and/or filament interacting like protein 3|FLJ12560|Nav 1|POMFIL 3
- Storage Buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage Temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping Temperature:4°C
- Purification:Purified by Protein A
- Cat. No.:10257-274
- Supplier no.:BS-11467R-A647
Specifications
About this item
Neuron navigator 1 is a 1877 amino acid cytoplasmic protein that is involved in neuronal migration. Neuron navigtor 1 is widely expressed at low levels, though highest expression is found in both adult and fetal nervous tissue. Through interaction with tubulin, Neuron navigator 1 associates with a subset of mirotubule plus ends present in the growth cone. Overexpression of Neuron navigator 1 leads to microtubule bundling, whereas a reduction of its levels causes loss of directionality in the migration of pontine cell leading processes. There are seven isoforms of Neuron navigator 1 that are produced as a result of alternative splicing events.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Neuron navigator 1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat