ContactUsLinkComponent
Gammex 851 Neoprene Surgical Gloves, Sterile, Ansell
Gammex 851 Neoprene Surgical Gloves, Sterile, Ansell
Supplier:  Ansell Healthcare
Certificates
undefined
Gammex 851 Neoprene Surgical Gloves, Sterile, Ansell
Supplier:  Ansell Healthcare
Specifications
  • Catalog No:
  • 94000-636
  • 94000-630
  • 94000-634
  • 94000-626
  • 94000-624
  • 94000-632
  • 94000-638
  • 94000-628
  • Ambidextrous/Hand-specific:
  • Hand-Specific
  • Hand-Specific
  • Hand-Specific
  • Hand-Specific
  • Hand-Specific
  • Hand-Specific
  • Hand-Specific
  • Hand-Specific
  • Disposable/Reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex/Latex-free:
  • Latex-Free
  • Latex-Free
  • Latex-Free
  • Latex-Free
  • Latex-Free
  • Latex-Free
  • Latex-Free
  • Latex-Free
  • Powdered/Powder-free:
  • Powder-Free
  • Powder-Free
  • Powder-Free
  • Powder-Free
  • Powder-Free
  • Powder-Free
  • Powder-Free
  • Powder-Free
  • Finish:
  • Smooth
  • Smooth
  • Smooth
  • Smooth
  • Smooth
  • Smooth
  • Smooth
  • Smooth
  • Cuff Style:
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Beaded
  • Cuff Thickness:
  • 6.3 mils
  • 6.3 mils
  • 6.3 mils
  • 6.3 mils
  • 6.3 mils
  • 6.3 mils
  • 6.3 mils
  • 6.3 mils
  • Finger Thickness:
  • 7.9 mils
  • 7.9 mils
  • 7.9 mils
  • 7.9 mils
  • 7.9 mils
  • 7.9 mils
  • 7.9 mils
  • 7.9 mils
  • Palm Thickness:
  • 7.5 mils
  • 7.5 mils
  • 7.5 mils
  • 7.5 mils
  • 7.5 mils
  • 7.5 mils
  • 7.5 mils
  • 7.5 mils
  • FDA Regulated:
  • +
  • +
  • +
  • +
  • +
  • +
  • +
  • +
Specifications