Specifications
- Description:MINI-SPIKE® Dispensing Pin with Security Clip
- Environmentally Preferable:
- Cat. No.:10017-778
- Supplier no.:412004
Specifications
About this item
The security clip enables attachment to standard 20 mm rubber-stoppered vials.
They are also used for preparing and dispensing diluent or additive from multi-dose vials. It utilizes a bacterial retentive air venting filter and a proximal luer lock connector.