ContactUsLinkComponent
VACUETTE® Blood Culture Holder, Greiner Bio-One
  10017-802
VACUETTE® Blood Culture Holder, Greiner Bio-One
  10017-802
 :  450181
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    VACUETTE® Blood Culture Holder
  • Environmentally Preferable:
  • Cat. No.:
    10017-802
  • Supplier no.:
    450181
The VACUETTE® Blood Culture Holder is used to fill blood into blood culture bottles and tubes in routine venipuncture procedures.
  • Single-use
  • Nonsterile
  • Plastic construction
  • Compatible with all VACUETTE® Luer adapters
This device is to be used by properly trained healthcare professionals only in accordance with these instructions.
The VACUETTE® Blood Culture Holder is single-use, nonsterile, and made out of plastic. It is compatible with all VACUETTE® Luer Adapters and VACUETTE® Safety/Blood Collection Sets.