ContactUsLinkComponent
Sub-X® Clearing Agent, Leica
Sub-X® Clearing Agent, Leica
Catalog # 10015-094
Supplier:  Leica Biosystems
CAS Number:  
undefined
Sub-X® Clearing Agent, Leica
Catalog # 10015-094
Supplier:  Leica Biosystems
Supplier Number:  3803670
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Sub-X® Clearing Agent
  • Size:
    1 gal.
  • Environmentally Preferable:
  • Cat. No.:
    10015-094
  • Supplier no.:
    3803670
Specifications
About this item
Sub-X® Clearing Agent is an economical xylene substitute that is an essentially odorless and non-greasy aliphatic hydrocarbon.
  • Economical
  • Odorless
  • Non-greasy aliphatic hydrocarbon
  • Much safer toxicity level than xylene
  • Safe to use with most plastics and tubing
It can be used in place of xylene for tissue processing, deparaffinization, clearing, and cleanup. Sub-X® is a gentle clearant which will not over-harden tissue specimens. The toxicity level is three times safer than xylene. This selective solvent is safe to use with most plastics and tubing in open and closed tissue processors.
A compatible mountant such as Sub-X® Mounting Medium must be used for coverslipping.