To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antigen Name:huntingtin interacting protein 1 related
- Antigen Symbol:HIP1R
- Clonality:Polyclonal
- Host:Multiple
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:1 Set
- Environmentally Preferable:
- Gene ID:9026
- Antigen Synonyms:ILWEQ|HIP12|HIP3
- Cat. No.:10539-052
- Supplier no.:H00009026-PW1
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-HIP1R (300 µl); 2. Antibody pair for Western Blot: Mouse purified Polyclonal Anti-HIP1R (50 µg)
Type:
Antigen: HIP1R
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human