Anti-KIF5B Rabbit Polyclonal Antibody
Catalog # RL100401J04S
Supplier: Rockland Immunochemical
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:KIF5B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:25 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Antigen Synonyms:Kinesin-5|KIF11
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide
- Storage Temperature:–20 °C
- Concentration:46 mg/ml by Refractometry
- Shipping Temperature:Dry Ice
- Immunogen:Eg5 was prepared from whole rabbit serum produced by repeated immunizations with a truncated Eg5 construct expressed in E. coli corresponding to human Eg5 protein.
- Purification:Anti-Eg5 is directed against the human Eg5 protein. The product was prepared from monospecific antiserum by delipidation and defibrination. A BLAST analysis was used to suggest reactivity with human. Cross-reactivity with Eg5 from other sources have not been determined.
- Cat. No.:RL100401J04S
- Supplier no.:100-401-J04S
Specifications
About this item
Type: Primary
Antigen: KIF5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human