Order Entry
United States
ContactUsLinkComponent
Anti-P30 and pro-caspases of p38 and p43 Rat Monoclonal Antibody [clone: Cas11.17D9]
Anti-P30 and pro-caspases of p38 and p43 Rat Monoclonal Antibody [clone: Cas11.17D9]
Supplier:  Biolegend
Certificates
Anti-P30 and pro-caspases of p38 and p43 Rat Monoclonal Antibody [clone: Cas11.17D9]
Supplier:  Biolegend
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 647202-BL
  • 647201-BL
  • Antigen Name:
  • P30 and pro-caspases of p38 and p43
  • P30 and pro-caspases of p38 and p43
  • Antigen Synonyms:
  • Casp11
  • Casp11
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Cas11.17D9
  • Cas11.17D9
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 201161
  • 201161
  • Isotype:
  • IgG2a
  • IgG2a
  • Western Blot:
  • Yes
  • Yes
  • Immunogen:
  • His tagged bacterial fusion protein of p 30 of caspase 11
  • His tagged bacterial fusion protein of p 30 of caspase 11
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Purification:
  • The antibody was purified by affinity chromatography.
  • The antibody was purified by affinity chromatography.
  • Concentration:
  • 0.50 mg/mL
  • 0.50 mg/mL
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

Specifications