About this item
Alexa Fluor® 647 anti-human FOXP3 [259D]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Rhesus, Baboon, Chimpanzee; Apps: ICFC; Size: 100 tests
Each lot of this antibody is quality control tested by intracellular flow cytometry using our True-Nuclear™ Transcription Factor Staining Protocol. For flow cytometric staining, the suggested use of this reagent is 5 μl per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: FOXP3
Clonality: Monoclonal
Clone: 259D
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Cynomolgus;Rhesus;Baboon;Chimpanzee
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 320213-BL
- 320214-BL
- Antigen Symbol:
- FOXP3
- FOXP3
- Antigen Synonyms:
- Zinc finger protein JM2|JM2|Forkhead box protein P3|Scurfin|IPEX
- Zinc finger protein JM2|JM2|Forkhead box protein P3|Scurfin|IPEX
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Alexa Fluor® 647
- Alexa Fluor® 647
- Clone:
- 259D
- 259D
- Reactivity:
- Baboon, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human
- Baboon, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 50943
- 50943
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Full-length FOXP3 protein
- Full-length FOXP3 protein
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with Alexa Fluor® 647 under optimal conditions.
- The antibody was purified by affinity chromatography, and conjugated with Alexa Fluor® 647 under optimal conditions.
- Storage Temperature:
- The FOXP3 antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The FOXP3 antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT