About this item
Alexa Fluor® 488 anti-mouse/human CD15 (SSEA-1) [MC-480]; Isotype: Mouse IgM, κ; Reactivity: Mouse, Cross-Reactivity: Human; Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: SSEA-1
Clonality: Monoclonal
Clone: MC-480
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: Mouse IgM, κ
Reactivity: Mouse;Cross-Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 125610-BL
- 125609-BL
- Antigen Symbol:
- SSEA-1
- SSEA-1
- Antigen Synonyms:
- CD15|stage-specific embryonic anti-gen-1
- CD15|stage-specific embryonic anti-gen-1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Alexa Fluor® 488
- Alexa Fluor® 488
- Clone:
- MC-480
- MC-480
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 14345, 2526
- 14345, 2526
- Isotype:
- IgM kappa
- IgM kappa
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Mouse F9 Teratocarcinoma Stem Cells (X-irradiated)
- Mouse F9 Teratocarcinoma Stem Cells (X-irradiated)
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Purification:
- The antibody was conjugated with Alexa Fluor® 488 under optimal conditions, and is at >85% purity.
- The antibody was conjugated with Alexa Fluor® 488 under optimal conditions, and is at >85% purity.
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT