Anti-CENPU Rabbit Polyclonal Antibody
RL600-401-A92
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Centromere protein U
- Antigen Symbol:CENPU
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Epitope:N-Terminal
- Cross Adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:79682
- Antigen Synonyms:MLF1IP|CENPU50|PBIP1|KLIP1|CENP50
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage Temperature:–20 °C
- Concentration:0.88 mg/ml by UV absorbance at 280 nm
- Shipping Temperature:Dry Ice
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a 200 residue recombinant Protein corresponding to the amino terminal end of human MLF1IP Protein.
- Purification:affinity-purified
- Cat. No.:RL600-401-A92
- Supplier no.:600-401-A92
Type: Primary
Antigen: CENPU
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: