Order Entry
United States
ContactUsLinkComponent
Anti-ACTIVE® JNK Rabbit Polyclonal Antibody (pTPpY)
Anti-ACTIVE® JNK Rabbit Polyclonal Antibody (pTPpY)
Catalog # PAV7931
Supplier:  Avantor
Anti-ACTIVE® JNK Rabbit Polyclonal Antibody (pTPpY)
Catalog # PAV7931
Supplier:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antigen Name:
    ACTIVE® JNK
  • Antigen Symbol:
    JNK
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
  • Size:
    40 μl
  • Environmentally Preferable:
  • Storage Temperature:
    –30 °C to –10 °C
  • Cat. No.:
    PAV7931
  • Supplier no.:
    V7931

Specifications

About this item

Anti-ACTIVE JNK pAb is a polyclonal rabbit antibody that recognizes the active form of JNK enzymes.

  • Preferentially Detects Active, Dually Phosphorylated JNK
  • Affinity-purified rabbit IgG
  • Use for Western blotting, immunocytochemistry and Immunohistochemistry

Anti-ACTIVE JNK pAb is a polyclonal antibody from rabbit serum. The antibody is affinity purified using a dually phosphorylated peptide that corresponds to the active form of the JNK enzymes.

Type:
Antigen: ACTIVE® JNK
Clonality: Polyclonal
Clone:
Conjugation: pTPpY
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: