To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:forkhead box H1
- Antigen Symbol:FOXH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.02 mg
- Environmentally Preferable:
- Epitope:near the N-Terminus
- Cross Adsorption:No
- Form:Liquid
- Gene ID:O75593
- Antigen Synonyms:FOXH1
- Storage Buffer:FOXH1 Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular Weight:40 kDa
- Storage Temperature:FOXH1 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Rabbit polyclonal FOXH1 antibody was raised against a 19 amino acid peptide near the amino terminus of human FOXH1. The immunogen is located within the first 50 amino acids of FOXH1.
- Purification:FOXH1 Antibody is affinity chromatography purified via peptide column.
- Cat. No.:10751-802
- Supplier no.:7095_S
Specifications
About this item
FOXH1 Antibody: FOXH1 is a member of the Forkhead-box (FOX) family of transcription factors and is the human homolog of Xenopus forkhead activin signal transducer-1. FOXH1 plays an important role in mediating TGF-beta/activin signals through its interaction with the SMAD2-SMAD4 complex. It is expressed primarily during early development and localizes in the cytoplasm until it forms a complex with the SMAD proteins at which time it translocates to the nucleus to positively or negatively affect gene expression.
Type: Primary
Antigen: FOXH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat