Order Entry
United States
ContactUsLinkComponent
Anti-PELI1 Rabbit Polyclonal Antibody
Anti-PELI1 Rabbit Polyclonal Antibody
Catalog # 10751-782
Supplier:  Prosci
undefined
Anti-PELI1 Rabbit Polyclonal Antibody
Catalog # 10751-782
Supplier:  Prosci
Supplier Number:  7071_S
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    pellino E3 ubiquitin protein ligase 1
  • Antigen Symbol:
    PELI1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    0.02 mg
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    Q96FA3
  • Antigen Synonyms:
    PELI1
  • Storage Buffer:
    Pellino 1 Antibody is supplied in PBS containing 0.02% sodium azide.
  • Molecular Weight:
    46 kDa
  • Storage Temperature:
    Pellino 1 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Concentration:
    1 mg/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Rabbit polyclonal Pellino 1 antibody was raised against a 15 amino acid peptide near the carboxy terminus of human Pellino 1. The immunogen is located within amino acids 300 - 350 of Pellino 1.
  • Tested Applications:
    Immunocytochemistry
  • Purification:
    Pellino 1 Antibody is affinity chromatography purified via peptide column.
  • Cat. No.:
    10751-782
  • Supplier no.:
    7071_S

Specifications

About this item

Pellino 1 Antibody: The Pellino proteins are a highly homologous family of E3 ubiquitin ligases that act as upstream mediators in Toll-like receptor (TLR) pathways that lead to activation of MAP kinases and transcription factors. Pellino 1 is required for interleukin-1-mediated signaling through its interaction with the IRAK4-IRAK-TRAF6 complex, ultimately resulting in the activation of NF-kappa B. Like other members of the Pellino family, Pellino 1 is an E3 ubiquitin ligase, able to catalyze the polyubiquitination of IRAK1. It is activated via phosphorylation by either IRAK1 and IRAK4 or the IKK-related kinases IKK-epsilon and TBK1. In addition to phosphorylation, Pellino 1 activity is also modulated via ubiquitination and sumoylation.

Type: Primary
Antigen: PELI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat