Order Entry
United States
ContactUsLinkComponent
Anti-ATG10 Rabbit Polyclonal Antibody
Anti-ATG10 Rabbit Polyclonal Antibody
Catalog # 10750-032
Supplier:  Prosci
undefined
Anti-ATG10 Rabbit Polyclonal Antibody
Catalog # 10750-032
Supplier:  Prosci
Supplier Number:  4399_S
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    ATG10 autophagy related 10 homolog (S. cerevisiae)
  • Antigen Symbol:
    ATG10
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    0.02 mg
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    Q9H0Y0
  • Antigen Synonyms:
    ATG10
  • Storage Buffer:
    ATG10 Antibody is supplied in PBS containing 0.02% sodium azide.
  • Storage Temperature:
    ATG10 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Concentration:
    1 mg/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    ATG10 antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human ATG10. The immunogen is located within the last 50 amino acids of ATG10.
  • Purification:
    ATG10 Antibody is affinity chromatography purified via peptide column.
  • Cat. No.:
    10750-032
  • Supplier no.:
    4399_S

Specifications

About this item

ATG10 Antibody: Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein APG1. Another member of the autophagy protein family is ATG10, an E2-like enzyme involved in two ubiquitin-like modifications essential for autophagosome formation: ATG12-ATG5 conjugation and the modification of a soluble form of MAP-LC3, a homolog of yeast Apg8, to a membrane-bound form. ATG10 has also been shown to interact with ATG12 in human embryonic kidney cells in the presence of ATG7. Multiple isoforms of ATG10 are known to exist.

Type: Primary
Antigen: ATG10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat