To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:StrepII-tag Antibody
- Antigen Symbol:Strep-Tag II
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:
- Western Blot:Yes
- Size:0.02 mg
- Environmentally Preferable:
- Cross Adsorption:No
- Form:Liquid
- Antigen Synonyms:Strep-Tag II,Strep-Tag IIantibody|anti-Strep-Tag II|Strep-Tag II
- Storage Buffer:StrepII-tag Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage Temperature:StrepII-tag antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:StrepII-tag antibody was raised against the amino acid sequence "srWSHPQFEKsh".
- Purification:StrepII-tag Antibody is affinity chromatography purified via peptide column.
- Cat. No.:10749-150
- Supplier no.:4335_S
StrepII-tag Antibody: Streptavidin is a tetrameric protein purified from Streptomyces avidinii. It has wide use in numerous molecular biological protocols dues to its strong affinity for biotin. The original Strep-tag is a nine amino acid peptide with high specificity and affinity towards streptavidin. The addition of this tag to the C-terminus of recombinant proteins allowed the simple purification of protein by use of affinity columns. It was re-engineered to allow it to also placed at the N-terminus of recombinant proteins.
Type: Primary
Antigen: StrepII-tag Antibody
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Tag