About this item
The high performance Elmasonic S ultrasonic baths operate in three ultrasonic modes to prepare samples, clean parts, and degas solvents. S line ultrasonic cleaners have a convenient LED display of set and actual temperature and time.
- Wide range of tank sizes
- All models have timer, heater, drain
- 3 ultrasonic modes optimized for mixing, cleaning, and degassing
- LED display of set and actual temperature and time
- Quiet operation
The Elmasonic S ultrasonic bath operates at 37 kHz in three ultrasonic modes: Normal, Sweep, and Degas. Samples are rapidly mixed, dispersed, dissolved, or homogenized in Normal mode. Sweep mode produces a continuous slight variation of frequency to provide uniform and thorough cleaning of parts. A series of LEDs continuously show set time, time remaining, set temperature, and actual temperature. No need to scroll through menus to view parameters. The microprocessor-controlled generator in this sonic cleaner maintains constant ultrasonic power, sensing and adjusting for light or heavy loads in the tank. Digital operation with power tracking ensures reproducible performance from run to run.
All Elma Ultrasonic S models have a rear drain with a valve on the side of the tank for easy access. Temperature is adjustable from 30°C to 80°C; the timer can be set from 1-30 minutes or continuous operation. The unit turns off automatically after 12 hours of continuous operation for safety. The tank cover can be flipped over and used as a drip tray.
Dependable, state-of-the-art Elma ultrasonic cleaners are made in Germany.