Microprocessor based pH 700 benchtop meter measures pH, mV, relative mV, and temperature.
- Large display permits easier readout viewing
- The large, dual-display LCD provides excellent visibility, even from distance
- Datalogging allows you to store and recall up to 100 data sets
- Automatic temperature compensation (ATC) maintains reading accuracy even with fluctuating temperatures
Five point pH calibration, with automatic buffer recognition for both USA (pH 1.68, 4.01, 7.00, 10.01, 12.45) and NIST (pH 1.68, 4.01, 6.86, 9.18, 12.45) pH buffers, makes for easy calibration no matter which requirement you need to meet.
Meter features HOLD, ATC and electrode diagnostics. The convenient pull-out quick reference card keeps procedures handy at all times. Meter accepts any pH or ORP electrodes with a BNC connector.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 101076-308
- 76221-212
- 101076-306
- pH Range:
- −2...+16
- −2...+16
- −2...+16
- pH Resolution:
- 0.01/0.1
- 0.01/0.1
- 0.01/0.1
- pH Accuracy:
- ±0.01
- ±0.01
- ±0.01
- pH Calibration:
- 5 point
- 5 point
- 5 point
- Temperature Range:
- 0.0...100.0 °C (32...212 °F)
- 0.0...100.0 °C (32...212 °F)
- 0.0...100.0 °C (32...212 °F)
- Temperature Compensation:
- Automatic or manual
- Automatic or manual
- Automatic or manual
- Display:
- Dual-display LCD
- Dual-display LCD
- Dual-display LCD
- Power supply:
- 110 - 220 V
- 110 - 220 V
- 110 - 220 V
- Data Logging Capacity:
- 100 data sets
- 100 data sets
- 100 data sets
- Dimensions:
- 15.5 x 17.5 x 6.9 cm (61/8 x 67/8 x 23/4)"
- 15.5 x 17.5 x 6.9 cm (61/8 x 67/8 x 23/4)"
- 15.5 x 17.5 x 6.9 cm (61/8 x 67/8 x 23/4)"