To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Serine Incorporator 1
- Antigen Symbol:SERC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:57515
- Antigen Synonyms:PLI|TMS2|A2AP|TDE2|TMS-2|ALPHA-2-PI|API|AAP|TDE1L
- Storage Buffer:Phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
- Storage Temperature:Store at -20 degrees Celcius. Product is stable for at least 1 year.
- Immunogen:The antiserum was produced against synthesized peptide derived from human SERC1.
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen.
- Cat. No.:89290-484
- Supplier no.:GTX87464
Specifications
About this item
Rabbit polyclonal antibody to SERC1
We recommend the following starting dilutions: WB: 1:500~1:1000 ELISA: 1:20000Optimal dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: SERC1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat