Order Entry
United States
ContactUsLinkComponent
Anti-ITGA1 Rabbit Polyclonal Antibody
Anti-ITGA1 Rabbit Polyclonal Antibody
  89301-118
 :  Genetex
undefined
Anti-ITGA1 Rabbit Polyclonal Antibody
  89301-118
 :  Genetex
 :  GTX81008
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Integrin Alpha 1
  • Antigen Symbol:
    ITGA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    400 μl
  • Environmentally Preferable:
  • Gene ID:
    3672
  • Antigen Synonyms:
    integrin alpha-1|ITGA1|laminin and collagen receptor|VLA-1|very late activation protein 1|CD49 antigen-like family member A
  • Storage Buffer:
    PBS with 0.09% (W/V) sodium azide
  • Storage Temperature:
    Store at at 2-8°C for up to 6 months. For long term storage, aliquot and store at -20 °C. Avoid repeated freeze/thaw cycles.
  • Concentration:
    0.25mg/ml
  • Immunogen:
    KLH conjugated synthetic peptide between 734~763 amino acids from the Center region of human ITGA1.
  • Purification:
    Purified through a protein A column, followed by peptide affinity purification.
  • Cat. No.:
    89301-118
  • Supplier no.:
    GTX81008

 

 

Rabbit Polyclonal antibody to Integrin alpha 1

Recommended Starting Dilutions:ELISA: Use at 1:1,000WB: Use at 1:100-500IHC: Use at 1:50-100Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: ITGA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse