To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Killer Cell Immunoglobulin-Like Receptor, Two Domains, Short Cytoplasmic Tail, 4
- Antigen Symbol:KIR Family
- Clonality:Monoclonal
- Clone:NKVFS1
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:3809
- Antigen Synonyms:NKAT8|KKA3|NKAT-8|KIR1D|CD158I|KIR412|KIR2DS1
- Storage Buffer:Phosphate buffered saline containing 0.09% Sodium Azide
- Storage Temperature:Store at +4 °C or at –20 °C if preferred. This product should be stored undiluted. Storage in frost-free freezers is not recommended. Should this product contain a precipitate we recommend microcentrifugation before use.
- Purification:Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
- Cat. No.:89277-404
- Supplier no.:GTX76004
Specifications
About this item
Mouse monoclonal antibody [NKVFS1] to KIR family
Recommended Starting Dilutions: Flow Cytometry: 1/50 - 1/100 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: KIR Family
Clonality: Monoclonal
Clone: NKVFS1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: