Order Entry
United States
ContactUsLinkComponent
 
Anti-CK14 Rabbit Monoclonal Antibody [clone: EPR1612]
  89327-376
 :  Genetex
undefined
Anti-CK14 Rabbit Monoclonal Antibody [clone: EPR1612]
  89327-376
 :  Genetex
 :  GTX62683
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Cytokeratin 14
  • Antigen Symbol:
    CK14
  • Clonality:
    Monoclonal
  • Clone:
    EPR1612
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    3861
  • Antigen Synonyms:
    Dowling-Meara|keratin 14 (epidermolysis bullosa simplex|type I cytoskeletal 14|cytokeratin-14|cytokeratin 14|keratin-14|CK-14|Koebner)|keratin
  • Storage Buffer:
    PBS containing 50% Glycerol, 0.05% BSA and 0.01% Sodium azide.
  • Storage Temperature:
    Store at –20 °C. Stable for 12 months at –20 °C
  • Immunogen:
    Synthetic peptide corresponding to residues in human Cytokeratin 14.
  • Purification:
    Tissue culture supernatant
  • Cat. No.:
    89327-376
  • Supplier no.:
    GTX62683

 

 

Rabbit monoclonal antibody [EPR1612] to Cytokeratin 14

Type: Primary
Antigen: CK14
Clonality: Monoclonal
Clone: EPR1612
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: