To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Tumer Necrosis Factor Receptor 1
- Antigen Symbol:TNFR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Hamster,Rabbit,Monkey,Human,Bovine,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:21937
- Antigen Synonyms:TNFRSF1A|Tumor necrosis factor receptor superfamily member 1A [Precursor]|TNFR-I|p55|CD120a antigen|TNFAR|p60|TNF-R1|TNFR1|TNF-RI
- Storage Buffer:PBS (pH 7.2) containing 30% glycerol, 0.5% BSA, 0.01% thimerosal
- Storage Temperature:The antibody solution should be gently mixed before use. Stored at -20 °C for future use.
- Concentration:0.2 mg/ml
- Purification:Antigen affinity purification
- Cat. No.:89279-834
- Supplier no.:GTX59625
Specifications
About this item
Rabbit polyclonal antibody to TNF-R1
Western blotting (0.5-4 µg/ml) and in Immunohistochemistry (10-20 µg/ml). However, the optimal conditions should be determined individually. The antibody detects cdc42 of human, mouse, rat, and bovine origins.
Type: Primary
Antigen: TNFR1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Hamster, Monkey, Rabbit, Rat, Bovine