To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Translocator protein
- Antigen Symbol:TSPO
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:2257
- Antigen Synonyms:BPBS|translocator protein|MBR|DBI|IBP|PBS|pk18|PTBR|BZRP|PKBS|mDRC|PBR
- Storage Buffer:Phosphate buffered saline (PBS), pH 7.2.
- Storage Temperature:The antibody solution should be gently mixed before use. Stored at -20 °C for future use.
- Concentration:0.2 mg/ml
- Purification:Antigen affinity purification
- Cat. No.:89271-070
- Supplier no.:GTX59606
Rabbit polyclonal antibody to PBR
Western blot analysis (0.5-4 µg/ml) and Immunohistochemistry (10-20 µg/ml). However, the optimal conditions should be determined individually.
Type: Primary
Antigen: TSPO
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat