Anti-GSK3B Rabbit Polyclonal Antibody
Catalog # 89300-342
Supplier: Genetex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Glycogen Synthase Kinase 3 beta
- Antigen Symbol:GSK3B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Zebrafish,Xenopus,Chicken,Human,Bovine,Dog,Rat,Mouse
- Western Blot:Yes
- Size:50 μl
- Environmentally Preferable:
- Gene ID:2932
- Antigen Synonyms:EC 2.7.11.26|GSK3B|Factor A|GSK-3 beta Factor A|GSK-3 beta
- Modification:phospho-Ser9
- Storage Buffer:10 mM Hepes (pH7.5), 150 mM NaCl, 100 ug/ml BSA
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:Phosphopeptide corresponding to amino acid residues surrounding the phospho- Ser9 of GSK3beta. Prepared from rabbit serum by affinity purification via sequential chromatography on phospho- and dephosphopeptide affinity columns.
- Purification:Antigen affinity purification
- Cat. No.:89300-342
- Supplier no.:GTX48612
Specifications
About this item
Rabbit polyclonal antibody to GSK3 alpha/beta (phospho Ser9)
Western Blot 1:1000
Type: Primary
Antigen: GSK3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat