To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Thymocyte-expressed Molecule Involved in Selection
- Antigen Symbol:THEMIS
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 μg
- Environmentally Preferable:
- Gene ID:387357
- Antigen Synonyms:thymocyte-expressed molecule involved in selection|THEMIS|bA325O24.3|FLJ40584|thymocyte selection associated|bA325O24.4|TSEPA|thymocyte selection pathway associated|C6orf190|MGC163388|C6orf207
- Storage Buffer:Phosphate, pH7.2-7.4
- Storage Temperature:Product should be aliquoted and stored at -20∞C. Avoid repeated freeze/thaw cycles.
- Immunogen:Synthetic peptide
- Purification:Affinity purified using antigen.
- Cat. No.:89275-880
- Supplier no.:GTX47850
FITC-conjugated Rabbit polyclonal antibody to Thymocyte selection associated protein
We recommend the following starting dilutions: 1:500 for WB 1:150 for IHC Optimal working dilutions should be determined experimentally be the end user.
Type: Primary
Antigen: THEMIS
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human