To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Centromere protein B, 80kDa
- Antigen Symbol:CENPB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 μg
- Environmentally Preferable:
- Gene ID:1059
- Antigen Synonyms:CENP B|Centromere protein B|CENP-B|centromere autoantigen B|Major centromere autoantigen B|CENPB_HUMAN.
- Storage Buffer:Phosphate buffered saline (PBS)
- Storage Temperature:Store at 4 °C for short-term use. For longer periods of storage, aliquot and store at –20 °C. Avoid repeat freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Synthetic peptide
- Purification:Peptide Affinity Purified
- Cat. No.:89268-062
- Supplier no.:GTX45813
Specifications
About this item
Rabbit polyclonal antibody to CENPB (C-terminal)
Recommended Dilutions: For Western Blot: Use at 1 ug/ml.
Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.
Type: Primary
Antigen: CENPB (centromere Protein B 80kDa)
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: