To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Adiponectin Receptor 2
- Antigen Symbol:ADIPOR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:79602
- Antigen Synonyms:ACDCR2|Adiponectin receptor protein 2|FLJ21432|Progestin and adipoQ receptor family member II|MGC4640|PAQR2
- Storage Buffer:Phosphate buffered saline containing 0.1% Sodium Azide (NaN3) 0.1% Bovine Serum Albumin
- Storage Temperature:Store at –20 °C only. Storage in frost-free freezers is not recommended. This product should be stored undiluted. Should this product contain a precipitate we recommend microcentrifugation before use.
- Immunogen:Synthetic peptide CSRTPEPDIRLRKGHQLDGTRRGDNDSHQG corresponding to amino acids 11-40 within the N-terminal region of human AdipoR2.
- Purification:Purified IgG was prepared by affinity chromatography.
- Cat. No.:89275-712
- Supplier no.:GTX42847
Goat polyclonal antibody to AdipoR2
Recommended Starting Dilutions: Immunohistology - Paraffin: Min Dilution 1/250 Western Blotting: Min Dilution 1/1000 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.
Type: Primary
Antigen: ADIPOR2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human