Anti-MST1R Rabbit Polyclonal Antibody
Catalog # 89384-178
Supplier: Abgent
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Macrophage Stimulating 1 Receptor
- Antigen Symbol:MST1R
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Antigen Synonyms:soluble R|RON variant E2E3|MST1R variant RON30|PTK8|macrophage-stimulating protein receptor|MST1R variant RON62|soluble RON variant 1|c-met-related tyrosine kinase|CDw136|PTK8 protein tyrosine kinase 8|MSP receptor|p185-Ron|soluble RON variant 3|RON|soluble RON variant 2|CD136
- Cat. No.:89384-178
- Supplier no.:AP14137B
Specifications
About this item
Type: Primary
Antigen: MST1R
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse